Assay Details
| Gene Symbol: LOC103347134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008253601.2 | XP_008251823.1 | 1535 | 100 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC103347134| Gene Symbol: LOC103347134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008253601.2 | XP_008251823.1 | 1535 | 100 | ||
Set Membership: |
|