Assay Details
| Gene Symbol: GATC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719791.3 | XP_002719837.3 | 2 - 3 | 277 | 71 | |
| XM_017349514.1 | XP_017205003.1 | 2 - 3 | 278 | 71 | ||
More Information
Set Membership: |
|

GATC
Genomic Map