Assay Details
| Gene Symbol: DNAJC7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719389.3 | XP_002719435.1 | 6 - 7 | 826 | 63 | |
| XM_008271484.2 | XP_008269706.2 | 6 - 7 | 541 | 63 | ||
More Information
Set Membership: |
|

DNAJC7
Genomic Map