Assay Details
| Gene Symbol: NAA50 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266924.2 | XP_008265146.1 | 4 - 5 | 470 | 109 | |
| XM_008266925.2 | XP_008265147.1 | 4 - 5 | 446 | 109 | ||
| XM_008266926.2 | XP_008265148.1 | 4 - 5 | 561 | 109 | ||
| XM_017347083.1 | XP_017202572.1 | 4 - 5 | 467 | 109 | ||
| XM_017347084.1 | XP_017202573.1 | 4 - 5 | 558 | 109 | ||
More Information
Set Membership: |
|

NAA50
Genomic Map