Assay Details
| Gene Symbol: CHST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716579.2 | XP_002716625.1 | 1 - 2 | 480 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002716579.2 transcripts.
Set Membership: |
|

CHST2
Genomic Map