Assay Details
| Gene Symbol: YWHAG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711904.3 | XP_002711950.1 | 1 | 180 | 123 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002711904.3 transcripts.
Set Membership: |
|

YWHAG
Genomic Map