Assay Details
| Gene Symbol: TMEM54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720669.3 | XP_002720715.1 | 586 | 62 | ||
| XM_017350264.1 | XP_017205753.1 | 605 | 62 | |||
| XM_017350266.1 | XP_017205755.1 | 508 | 62 | |||
More Information
Set Membership: |
|

TMEM54
Genomic Map