Assay Details
| Gene Symbol: MTFR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263555.2 | XP_008261777.2 | 8 | 2394 | 94 | |
| XM_017345384.1 | XP_017200873.1 | 8 | 2398 | 94 | ||
More Information
Set Membership: |
|

MTFR2
Genomic Map