Assay Details
| Gene Symbol: ZNF385B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712320.3 | XP_002712366.1 | 9 - 10 | 1552 | 58 | |
| XM_008258797.2 | XP_008257019.1 | 8 - 9 | 1170 | 58 | ||
| XM_008258798.2 | XP_008257020.1 | 8 - 9 | 1162 | 58 | ||
| XM_008258799.2 | XP_008257021.1 | 8 - 9 | 1265 | 58 | ||
| XM_008258801.2 | XP_008257023.1 | 8 - 9 | 1087 | 58 | ||
More Information
Set Membership: |
|

ZNF385B
Genomic Map