Assay Details
| Gene Symbol: LRR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718227.3 | XP_002718273.1 | 1 - 2 | 299 | 92 | |
| XM_008269622.2 | XP_008267844.1 | 1 - 2 | 299 | 92 | ||
More Information
Set Membership: |
|

LRR1
Genomic Map