Assay Details
| Gene Symbol: BABAM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709945.3 | XP_002709991.1 | 1 - 2 | 109 | 93 | |
| XM_017340849.1 | XP_017196338.1 | 1 - 2 | 111 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_002709945.3, XM_017340849.1 transcripts.
Set Membership: |
|

BABAM2
Genomic Map