Assay Details
Gene Symbol: TLCD3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002718891.3 | XP_002718937.1 | 1 - 2 | 305 | 55 | |
XM_017348912.1 | XP_017204401.1 | 1 - 2 | 365 | 55 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
