Assay Details
| Gene Symbol: PPM1G | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709952.3 | XP_002709998.1 | 1 - 2 | 333 | 57 | |
| XM_017340854.1 | XP_017196343.1 | 1 - 2 | 333 | 57 | ||
More Information
Set Membership: |
|

PPM1G
Genomic Map