Assay Details
| Gene Symbol: CASC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719352.3 | XP_002719398.1 | 7 - 8 | 1462 | 67 | |
| XM_008271432.2 | XP_008269654.1 | 7 - 8 | 1539 | 67 | ||
More Information
Set Membership: |
|

CASC3
Genomic Map