Assay Details
Gene Symbol: LOC100354991 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017337726.1 | XP_017193215.1 | 321 | 82 |

More Information
Set Membership: |
![]() ![]() |
Panther Classification:
