Assay Details
| Gene Symbol: UFM1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721100.3 | XP_002721146.1 | 1 | 246 | 72 | |
| XM_017350618.1 | XP_017206107.1 | 1 | 409 | 72 | ||
More Information
Set Membership: |
|

UFM1
Genomic Map