Assay Details
| Gene Symbol: TC2N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349433.1 | XP_017204922.1 | 3 - 4 | 647 | 62 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TC2N| Gene Symbol: TC2N | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017349433.1 | XP_017204922.1 | 3 - 4 | 647 | 62 | |
Set Membership: |
|