Assay Details
| Gene Symbol: PKIB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263450.2 | XP_008261672.1 | 2 - 3 | 315 | 84 | |
| XM_008263451.2 | XP_008261673.1 | 2 - 3 | 237 | 84 | ||
| XM_008263452.2 | XP_008261674.1 | 2 - 3 | 244 | 84 | ||
| XM_017345342.1 | XP_017200831.1 | 3 - 4 | 402 | 84 | ||
| XM_017345343.1 | XP_017200832.1 | 3 - 4 | 342 | 84 | ||
More Information
Set Membership: |
|

PKIB
Genomic Map