Assay Details
Gene Symbol: GPR88 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008264818.2 | XP_008263040.1 | 2 | 3838 | 90 | |
XM_008264820.2 | XP_008263042.1 | 2 | 3834 | 90 | ||
XM_008264821.2 | XP_008263043.1 | 2 | 3780 | 90 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
