Assay Details
| Gene Symbol: RASL10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719243.3 | XP_002719289.1 | 3 - 4 | 720 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RASL10B| Gene Symbol: RASL10B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719243.3 | XP_002719289.1 | 3 - 4 | 720 | 73 | |
Set Membership: |
|