Assay Details
| Gene Symbol: PKMYT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008250623.2 | XP_008248845.2 | 1337 | 59 | ||
| XM_008250624.2 | XP_008248846.1 | 1267 | 59 | |||
| XM_008250625.1 | XP_008248847.1 | 1326 | 59 | |||
| XM_017338743.1 | XP_017194232.1 | 1349 | 59 | |||
More Information
Set Membership: |
|

PKMYT1
Genomic Map