Assay Details
| Gene Symbol: ERI2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711810.3 | XP_002711856.1 | 7 - 8 | 733 | 78 | |
| XM_008257830.2 | XP_008256052.1 | 7 - 8 | 635 | 78 | ||
More Information
Set Membership: |
|

ERI2
Genomic Map