Assay Details
| Gene Symbol: LOC100341358 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262769.2 | XP_008260991.1 | 2 - 3 | 396 | 93 | |
| XM_008262770.2 | XP_008260992.1 | 2 - 3 | 396 | 93 | ||
| XM_008262771.2 | XP_008260993.1 | 2 - 3 | 396 | 93 | ||
| XM_017345042.1 | XP_017200531.1 | 2 - 3 | 396 | 93 | ||
More Information
Set Membership: |
|

LOC100341358
Genomic Map