Assay Details
| Gene Symbol: IRF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002721631.3 | XP_002721677.1 | 309 | 83 | ||
| XM_008248487.2 | XP_008246709.1 | 309 | 83 | |||
| XM_008248489.2 | XP_008246711.1 | 309 | 83 | |||
| XM_017337683.1 | XP_017193172.1 | 309 | 83 | |||
More Information
Set Membership: |
|

IRF4
Genomic Map