Assay Details
| Gene Symbol: MSH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002709880.3 | XP_002709926.1 | 2 - 3 | 588 | 66 | |
| XM_008254436.2 | XP_008252658.1 | 2 - 3 | 562 | 66 | ||
More Information
Set Membership: |
|

MSH6
Genomic Map