Assay Details
| Gene Symbol: ENTPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008270246.2 | XP_008268468.1 | 4 - 5 | 850 | 71 | |
| XM_008270248.2 | XP_008268470.1 | 4 - 5 | 643 | 71 | ||
| XM_008270249.2 | XP_008268471.1 | 4 - 5 | 482 | 71 | ||
More Information
Set Membership: |
|

ENTPD1
Genomic Map