Assay Details
| Gene Symbol: ZNF445 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008260356.2 | XP_008258578.1 | 3 - 4 | 819 | 65 | |
| XM_008260357.2 | XP_008258579.1 | 3 - 4 | 835 | 65 | ||
| XM_017343724.1 | XP_017199213.1 | 3 - 4 | 931 | 65 | ||
More Information
Set Membership: |
|

ZNF445
Genomic Map