Assay Details
| Gene Symbol: DGCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722749.3 | XP_002722795.1 | 3496 | 54 | ||
| XM_008250577.2 | XP_008248799.1 | 3502 | 54 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

DGCR8| Gene Symbol: DGCR8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002722749.3 | XP_002722795.1 | 3496 | 54 | ||
| XM_008250577.2 | XP_008248799.1 | 3502 | 54 | |||
Set Membership: |
|