Assay Details
| Gene Symbol: MPHOSPH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711659.3 | XP_002711705.1 | 1 - 2 | 58 | 81 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MPHOSPH6| Gene Symbol: MPHOSPH6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002711659.3 | XP_002711705.1 | 1 - 2 | 58 | 81 | |
Set Membership: |
|