Assay Details
| Gene Symbol: NAB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712343.3 | XP_002712389.1 | 4 - 5 | 1189 | 79 | |
| XM_017342991.1 | XP_017198480.1 | 4 - 5 | 1120 | 79 | ||
| XM_017342992.1 | XP_017198481.1 | 4 - 5 | 1189 | 79 | ||
More Information
Set Membership: |
|

NAB1
Genomic Map