Assay Details
| Gene Symbol: STRADB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002712520.3 | XP_002712566.1 | 4 - 5 | 533 | 71 | |
| XM_008258963.2 | XP_008257185.1 | 4 - 5 | 536 | 71 | ||
More Information
Set Membership: |
|

STRADB
Genomic Map