Assay Details
| Gene Symbol: MMD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002719282.3 | XP_002719328.1 | 2 - 3 | 375 | 60 | |
| XM_008271269.2 | XP_008269491.1 | 2 - 3 | 375 | 60 | ||
More Information
Set Membership: |
|

MMD
Genomic Map