Assay Details
| Gene Symbol: SPATA17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717401.3 | XP_002717447.2 | 6 - 7 | 712 | 76 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SPATA17| Gene Symbol: SPATA17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717401.3 | XP_002717447.2 | 6 - 7 | 712 | 76 | |
Set Membership: |
|