Assay Details
| Gene Symbol: PHACTR4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008265865.2 | XP_008264087.1 | 9 - 10 | 1836 | 64 | |
| XM_017346543.1 | XP_017202032.1 | 8 - 9 | 2051 | 64 | ||
| XM_017346544.1 | XP_017202033.1 | 8 - 9 | 1831 | 64 | ||
| XM_017346545.1 | XP_017202034.1 | 9 - 10 | 1837 | 64 | ||
More Information
Set Membership: |
|

PHACTR4
Genomic Map