Assay Details
| Gene Symbol: RSPO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720657.2 | XP_002720703.1 | 1365 | 57 | ||
| XM_008273651.1 | XP_008271873.1 | 1401 | 57 | |||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

RSPO1| Gene Symbol: RSPO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002720657.2 | XP_002720703.1 | 1365 | 57 | ||
| XM_008273651.1 | XP_008271873.1 | 1401 | 57 | |||
Set Membership: |
|