Assay Details
| Gene Symbol: RIPOR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008262486.2 | XP_008260708.1 | 14 - 15 | 2046 | 92 | |
| XM_008262488.2 | XP_008260710.1 | 14 - 15 | 2181 | 92 | ||
| XM_017344960.1 | XP_017200449.1 | 13 - 14 | 1893 | 92 | ||
More Information
Set Membership: |
|

RIPOR2
Genomic Map