Assay Details
| Gene Symbol: AP1AR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002717088.3 | XP_002717134.1 | 9 - 10 | 667 | 70 | |
| XM_002717089.3 | XP_002717135.1 | 8 - 9 | 568 | 70 | ||
More Information
Set Membership: |
|

AP1AR
Genomic Map