Assay Details
| Gene Symbol: HSPA13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002716764.3 | XP_002716810.1 | 2 - 3 | 591 | 75 | |
| XM_008267133.1 | XP_008265355.1 | 2 - 3 | 448 | 75 | ||
More Information
Set Membership: |
|

HSPA13
Genomic Map