Assay Details
| Gene Symbol: RUNX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008274944.2 | XP_008273166.1 | 307 | 69 | ||
| XM_008274946.2 | XP_008273168.1 | 308 | 69 | |||
| XM_017350826.1 | XP_017206315.1 | 313 | 69 | |||
| XM_017350827.1 | XP_017206316.1 | 316 | 69 | |||
More Information
Set Membership: |
|

RUNX1
Genomic Map