Assay Details
| Gene Symbol: STX11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008263598.2 | XP_008261820.1 | 1 - 2 | 87 | 98 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008263598.2 transcripts.
Set Membership: |
|

STX11
Genomic Map