Assay Details
| Gene Symbol: FYB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002713989.3 | XP_002714035.2 | 17 - 18 | 2560 | 87 | |
| XM_008262138.2 | XP_008260360.1 | 18 - 19 | 2700 | 87 | ||
| XM_008262139.2 | XP_008260361.1 | 18 - 19 | 2649 | 87 | ||
| XM_008262140.2 | XP_008260362.1 | 18 - 19 | 2607 | 87 | ||
| XM_008262141.2 | XP_008260363.1 | 18 - 19 | 2579 | 87 | ||
| XM_008262142.2 | XP_008260364.1 | 18 - 19 | 2685 | 87 | ||
More Information
Set Membership: |
|

FYB1
Genomic Map