Assay Details
| Gene Symbol: AP5M1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002718266.3 | XP_002718312.1 | 2 - 3 | 942 | 67 | |
| XM_017348333.1 | XP_017203822.1 | 2 - 3 | 942 | 67 | ||
More Information
Set Membership: |
|

AP5M1
Genomic Map