Assay Details
| Gene Symbol: RFESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344610.1 | XP_017200099.1 | 1 - 2 | 122 | 80 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_017344610.1 transcripts.
Set Membership: |
|

RFESD
Genomic Map