Assay Details
| Gene Symbol: RFESD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017344609.1 | XP_017200098.1 | 2 - 3 | 304 | 69 | |
| XM_017344610.1 | XP_017200099.1 | 2 - 3 | 298 | 69 | ||
| XM_017344611.1 | XP_017200100.1 | 2 - 3 | 558 | 69 | ||
More Information
Set Membership: |
|

RFESD
Genomic Map