Assay Details
| Gene Symbol: C12H6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714333.3 | XP_002714379.2 | 5 - 6 | 1079 | 73 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C12H6orf136| Gene Symbol: C12H6orf136 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714333.3 | XP_002714379.2 | 5 - 6 | 1079 | 73 | |
Set Membership: |
|