Assay Details
| Gene Symbol: LMBR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002715020.3 | XP_002715066.1 | 11 - 12 | 1106 | 64 | |
| XM_017345650.1 | XP_017201139.1 | 11 - 12 | 1106 | 64 | ||
| XM_017345651.1 | XP_017201140.1 | 11 - 12 | 1106 | 64 | ||
More Information
Set Membership: |
|

LMBR1
Genomic Map