Assay Details
| Gene Symbol: GNE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008266450.2 | XP_008264672.1 | 5 - 6 | 1224 | 60 | |
| XM_008266504.2 | XP_008264726.1 | 5 - 6 | 1118 | 60 | ||
More Information
Set Membership: |
|

GNE
Genomic Map