Assay Details
| Gene Symbol: TENT5A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002714547.3 | XP_002714593.1 | 2 - 3 | 937 | 100 | |
| XM_008263145.2 | XP_008261367.1 | 2 - 3 | 881 | 100 | ||
More Information
Set Membership: |
|

TENT5A
Genomic Map