Assay Details
| Gene Symbol: CCNJL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002710350.3 | XP_002710396.1 | 4 - 5 | 802 | 76 | |
| XM_008255345.2 | XP_008253567.1 | 3 - 4 | 619 | 76 | ||
More Information
Set Membership: |
|

CCNJL
Genomic Map