Assay Details
Gene Symbol: ODF3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008252743.2 | XP_008250965.2 | 1368 | 50 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
